Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313356.3 | NP_001300285.1 | 5 - 6 | 1411 | 66 | |
NM_001313357.3 | NP_001300286.1 | 4 - 5 | 1140 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |