Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srab-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313312.1 | NP_001300241.1 | 2 - 3 | 299 | 75 | |
NM_071154.2 | NP_503555.2 | 2 - 3 | 314 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |