Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071370.5 | NP_503771.1 | 2 - 3 | 371 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |