Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330807.1 | NP_001317839.1 | 10 - 11 | 1698 | 122 | |
NM_001330808.1 | NP_001317840.1 | 3 - 4 | 723 | 122 | ||
NM_071640.2 | NP_504041.1 | 10 - 11 | 1692 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |