Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srx-64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330820.1 | NP_001317762.1 | 5 - 6 | 798 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |