Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: taf-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071859.5 | NP_504260.1 | 2 - 3 | 153 | 62 | |
NM_071860.6 | NP_504261.1 | 2 - 3 | 138 | 62 |
Gene Symbol: K03B4.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380636.2 | NP_001367844.1 | 2 - 3 | 148 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |