Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071866.5 | NP_504267.3 | 8 - 9 | 1675 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |