Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srt-67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072004.3 | NP_504405.1 | 4 - 5 | 1474 | 91 | |
NR_136345.1 | 4 - 5 | 566 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |