Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129376.4 | NP_001122848.1 | 8 - 9 | 1611 | 61 | |
NM_072093.7 | NP_504494.2 | 7 - 8 | 1320 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |