Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dbl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072308.8 | NP_504709.1 | 5 - 6 | 1007 | 93 | |
GenBank mRNA | AF004395.1 | - | 5 - 6 | 965 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |