Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356677.4 | NP_001343695.1 | 20 | 7929 | 141 | |
NM_001356678.4 | NP_001343810.1 | 20 | 7929 | 141 | ||
NM_001356679.3 | NP_001343688.1 | 20 - 21 | 8018 | 141 | ||
NM_001356680.4 | NP_001343808.1 | 20 | 7929 | 141 | ||
NM_001356685.3 | NP_001343814.1 | 20 - 21 | 7923 | 141 | ||
NM_001356686.3 | NP_001343818.1 | 20 - 21 | 7923 | 141 | ||
NM_001356687.3 | NP_001343812.1 | 20 - 21 | 7923 | 141 | ||
NM_001356688.3 | NP_001343816.1 | 20 - 21 | 7923 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |