Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356673.4 | NP_001343696.1 | 42 | 13823 | 108 | |
NM_001356674.3 | NP_001343811.1 | 42 | 13826 | 108 | ||
NM_001356675.3 | NP_001343689.1 | 43 | 13865 | 108 | ||
NM_001356676.3 | NP_001343809.1 | 43 | 13868 | 108 | ||
NM_001356677.4 | NP_001343695.1 | 44 | 14171 | 108 | ||
NM_001356678.4 | NP_001343810.1 | 44 | 14174 | 108 | ||
NM_001356679.3 | NP_001343688.1 | 45 - 46 | 14302 | 108 | ||
NM_001356680.4 | NP_001343808.1 | 45 | 14216 | 108 | ||
NM_001356681.3 | NP_001343815.1 | 42 - 43 | 13817 | 108 | ||
NM_001356682.3 | NP_001343819.1 | 42 - 43 | 13820 | 108 | ||
NM_001356683.3 | NP_001343813.1 | 43 - 44 | 13859 | 108 | ||
NM_001356684.3 | NP_001343817.1 | 43 - 44 | 13862 | 108 | ||
NM_001356685.3 | NP_001343814.1 | 44 - 45 | 14165 | 108 | ||
NM_001356686.3 | NP_001343818.1 | 44 - 45 | 14168 | 108 | ||
NM_001356687.3 | NP_001343812.1 | 45 - 46 | 14207 | 108 | ||
NM_001356688.3 | NP_001343816.1 | 45 - 46 | 14210 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |