Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356673.4 | NP_001343696.1 | 8 | 3946 | 98 | |
NM_001356674.3 | NP_001343811.1 | 8 | 3946 | 98 | ||
NM_001356675.3 | NP_001343689.1 | 8 | 3946 | 98 | ||
NM_001356676.3 | NP_001343809.1 | 8 | 3946 | 98 | ||
NM_001356677.4 | NP_001343695.1 | 8 | 3946 | 98 | ||
NM_001356678.4 | NP_001343810.1 | 8 | 3946 | 98 | ||
NM_001356679.3 | NP_001343688.1 | 8 - 9 | 4035 | 98 | ||
NM_001356680.4 | NP_001343808.1 | 8 | 3946 | 98 | ||
NM_001356681.3 | NP_001343815.1 | 8 - 9 | 3940 | 98 | ||
NM_001356682.3 | NP_001343819.1 | 8 - 9 | 3940 | 98 | ||
NM_001356683.3 | NP_001343813.1 | 8 - 9 | 3940 | 98 | ||
NM_001356684.3 | NP_001343817.1 | 8 - 9 | 3940 | 98 | ||
NM_001356685.3 | NP_001343814.1 | 8 - 9 | 3940 | 98 | ||
NM_001356686.3 | NP_001343818.1 | 8 - 9 | 3940 | 98 | ||
NM_001356687.3 | NP_001343812.1 | 8 - 9 | 3940 | 98 | ||
NM_001356688.3 | NP_001343816.1 | 8 - 9 | 3940 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |