Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072411.1 | NP_504812.1 | 4 - 5 | 1164 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |