Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cri-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072712.8 | NP_505113.1 | 2 - 3 | 432 | 105 | |
GenBank mRNA | AF303253.1 | - | 2 - 3 | 432 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |