Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072932.4 | NP_505333.1 | 2 - 3 | 348 | 167 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |