Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322582.2 | NP_001309498.1 | 5 - 6 | 902 | 162 | |
NR_136369.1 | 5 - 6 | 878 | 162 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |