Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sru-40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269210.2 | NP_001256139.1 | 3 - 4 | 701 | 154 | |
NM_001269211.4 | NP_001256140.1 | 1 - 2 | 168 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |