Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: che-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269275.3 | NP_001256204.1 | 19 - 20 | 3104 | 66 | |
NM_001269276.3 | NP_001256205.1 | 10 - 11 | 1489 | 66 | ||
NM_001269277.3 | NP_001256206.1 | 8 - 9 | 1032 | 66 | ||
NM_001307803.4 | NP_001294732.1 | 8 - 9 | 1072 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |