Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373099.3 | NP_001360575.1 | 7 - 8 | 1158 | 101 | |
NM_073321.6 | NP_505722.1 | 7 - 8 | 1160 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |