Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sqv-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073329.10 | NP_505730.1 | 1 - 2 | 183 | 77 | |
GenBank mRNA | AY147932.1 | - | 1 - 2 | 185 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |