Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083273.4 | NP_001076742.1 | 5 - 6 | 1436 | 92 | |
NM_001083274.3 | NP_001076743.1 | 2 - 3 | 327 | 92 | ||
NM_001083275.4 | NP_001076744.1 | 4 - 5 | 531 | 92 | ||
NM_001356738.3 | NP_001343841.1 | 6 - 7 | 819 | 92 | ||
NM_001356739.4 | NP_001343842.1 | 4 | 541 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |