Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269388.2 | NP_001256317.1 | 2 - 3 | 94 | 73 | |
NM_001269389.2 | NP_001256318.1 | 2 - 3 | 94 | 73 | ||
GenBank mRNA | AY190131.1 | - | 1 - 2 | 45 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |