Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sas-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269409.2 | NP_001256338.1 | 3 - 4 | 603 | 102 | |
NM_001269410.3 | NP_001256339.1 | 1 - 2 | 184 | 102 | ||
NR_069564.1 | 3 - 4 | 527 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |