Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lon-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073660.6 | NP_506061.1 | 1 - 2 | 924 | 87 | |
GenBank mRNA | AF262406.1 | - | 1 - 2 | 880 | 87 | |
AF465981.1 | - | 1 - 2 | 884 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |