Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ent-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074120.4 | NP_506521.2 | 14 - 15 | 1937 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |