Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074186.7 | NP_506587.2 | 5 - 6 | 718 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |