Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ret-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129554.5 | NP_001123026.1 | 3 - 4 | 8042 | 66 | |
NM_001380820.1 | NP_001366847.1 | 1 - 2 | 120 | 66 | ||
NM_001383456.1 | 3 - 4 | 662 | 66 | |||
NM_074255.7 | NP_506656.2 | 3 - 4 | 8143 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |