Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srw-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074368.4 | NP_506769.2 | 3 - 4 | 2080 | 88 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |