Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srw-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074379.2 | NP_506780.2 | 1 - 2 | 524 | 166 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |