Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269742.2 | NP_001256671.1 | 5 - 6 | 672 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |