Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074700.1 | NP_507101.1 | 6 - 7 | 1039 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |