Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074832.3 | NP_507233.2 | 2 - 3 | 315 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |