Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028779.3 | NP_001023950.1 | 5 - 6 | 752 | 59 | |
NM_001028780.3 | NP_001023951.1 | 5 - 6 | 709 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |