Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322605.2 | NP_001309463.1 | 10 - 11 | 1591 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |