Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: par-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380857.1 | NP_001366977.1 | 4 - 5 | 494 | 153 | |
GenBank mRNA | AF160189.1 | - | 4 - 5 | 517 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |