Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270018.2 | NP_001256947.1 | 1 - 2 | 67 | 77 | |
NM_001373209.3 | NP_001360578.1 | 1 - 2 | 74 | 77 | ||
GenBank mRNA | AF293972.1 | - | 1 - 2 | 87 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |