Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028668.4 | NP_001023839.2 | 8 - 9 | 1528 | 100 | |
NM_001028669.4 | NP_001023840.2 | 8 - 9 | 1781 | 100 | ||
NM_001136423.4 | NP_001129895.2 | 8 - 9 | 1474 | 100 | ||
NM_001270032.3 | NP_001256961.1 | 8 - 9 | 1528 | 100 | ||
NM_001270033.2 | NP_001256962.1 | 8 - 9 | 1528 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |