Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075706.6 | NP_508107.1 | 4 - 5 | 361 | 122 | |
GenBank mRNA | U85766.1 | - | 4 - 5 | 351 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |