Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075706.6 | NP_508107.1 | 5 - 6 | 456 | 101 | |
GenBank mRNA | U85766.1 | - | 5 - 6 | 446 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |