Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: irk-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270047.4 | NP_001256976.2 | 1 - 2 | 106 | 132 | |
NM_001270048.3 | NP_001256977.1 | 1 - 2 | 114 | 132 | ||
NM_001307921.4 | NP_001294850.1 | 1 - 2 | 116 | 132 | ||
NM_001330888.3 | NP_001317907.1 | 2 - 3 | 205 | 132 | ||
NM_001380895.1 | NP_001368610.1 | 2 - 3 | 205 | 132 | ||
NM_001380896.1 | NP_001368348.1 | 2 - 3 | 225 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |