Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029433.7 | NP_001024604.1 | 5 - 6 | 806 | 78 | |
NM_001029434.5 | NP_001024605.1 | 3 - 4 | 578 | 78 | ||
NM_001313412.4 | NP_001300341.1 | 6 | 861 | 78 | ||
NM_001313413.4 | NP_001300342.1 | 5 - 6 | 808 | 78 | ||
NM_001313414.3 | NP_001300343.1 | 5 - 6 | 808 | 78 | ||
NM_001313415.4 | NP_001300344.1 | 3 - 4 | 525 | 78 | ||
NM_001380898.1 | NP_001368578.1 | 3 - 4 | 525 | 78 | ||
NM_075760.6 | NP_508161.3 | 6 - 7 | 910 | 78 | ||
GenBank mRNA | AF005205.1 | - | 3 - 4 | 573 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |