Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373217.4 | NP_001360743.1 | 12 - 13 | 1723 | 86 | |
NM_001380899.1 | NP_001367822.1 | 12 - 13 | 1717 | 86 | ||
NM_075777.7 | NP_508178.2 | 12 - 13 | 1683 | 86 | ||
NM_075778.5 | NP_508179.2 | 12 - 13 | 1724 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |