Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373217.4 | NP_001360743.1 | 11 - 12 | 1359 | 68 | |
NM_001380899.1 | NP_001367822.1 | 11 - 12 | 1353 | 68 | ||
NM_075778.5 | NP_508179.2 | 11 - 12 | 1360 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |