Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rme-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383525.1 | 4 - 5 | 398 | 125 | ||
NM_075807.5 | NP_508208.2 | 4 - 5 | 392 | 125 | ||
GenBank mRNA | AY750054.1 | - | 4 - 5 | 392 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |