Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029717.2 | NP_001024888.1 | 10 - 11 | 1470 | 91 | |
NM_001029718.2 | NP_001024889.1 | 13 - 14 | 1794 | 91 | ||
NM_001029719.1 | NP_001024890.1 | 11 - 12 | 1422 | 91 | ||
NM_001330893.1 | NP_001317860.1 | 12 - 13 | 1683 | 91 | ||
GenBank mRNA | AY275182.1 | - | 13 - 14 | 1794 | 91 | |
AY275183.1 | - | 11 - 12 | 1422 | 91 | ||
AY275184.1 | - | 10 - 11 | 1470 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |