Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vang-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076099.5 | NP_508500.1 | 5 - 6 | 1099 | 98 | |
NM_076100.7 | NP_508501.1 | 5 - 6 | 1076 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |