Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nmy-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001374981.3 | NP_001362097.1 | 16 - 17 | 5763 | 96 | |
NM_076103.6 | NP_508504.2 | 17 - 18 | 5785 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |