Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076121.7 | NP_508522.2 | 7 - 8 | 1315 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |