Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ddr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076171.6 | NP_508572.1 | 8 - 9 | 1453 | 105 | |
NM_076172.8 | NP_508573.1 | 8 - 9 | 1459 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |